Adam Equipment BKT Industrial Bench Scales with Built-In Label Printer
Get precise weights, input data easily and create custom labels
  • Ideal for manufacturing, shipping and receiving, packing and distribution warehouses
  • AE 503 indicator prints labels with pictures, logos, bar codes and QR codes
  • Level guide and adjustable feet ensure proper setup and optimum results
  • Enter data easily with color-coded keys
  • Capacity ranges up to 1320 lb (600 kg) across various models
Rugged construction well-suits industrial use, including weighing, parts counting, percentage weighing, checkweighing, animal weighing, dynamic weighing, averaging and other applications. Checkweighing features an audible alarm. Zero-tracking ensures the display returns to zero reading.
The easily visible backlit LCD displays 34-mm high digits, and the light adjusts per user preferences (always on, always off or on only when weighing). Included rechargeable battery aids convenience of placement and use; programmable auto power-off saves energy.
Printer supports multiple languages as it produces 50 mm-wide labels that can include bar codes, QR codes, pictures and custom logos. RS-232 interface lets users export data to computers and external printers.
268,000.00 - ₹443,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (lbs) Readability (g)
Availability Pricing
Adam Equipment BKT16a Industrial Bench Scale with Built-In Label Printer, 16lb/8kg, 11.8" x 15.7" Platform 1113000 Mfr # BKT 16A
COMPARE
External 16 0.2
₹280,000.00INR / Each
Adam Equipment BKT 35A Industrial Bench Scale with Built-In Label Printer, 35lb/6kg, 11.8" x 15.7" Platform 1113001 Mfr # BKT 35A
COMPARE
External calibration 35 0.5
Call For Price
Adam Equipment BKT 65A Industrial Bench Scale with Built-In Label Printer, 65lb/30kg, 11.8" x 15.7" Platform 1113002 Mfr # BKT 65A
COMPARE
External calibration 65 1
Call For Price
Adam Equipment BKT130a Industrial Bench Scale with Built-In Label Printer, 130lb/60kg, 11.8" x 15.7" Platform 1113003 Mfr # BKT 130A
COMPARE
External 130 2
₹268,000.00INR / Each
Adam Equipment BKT 260A Industrial Bench Scale with Built-In Label Printer, 260lb/120kg, 11.8" x 15.7" Platform 1113004 Mfr # BKT 260A
COMPARE
External calibration 260 5
Call For Price
Adam Equipment BKT165a Industrial Bench Scale with Built-In Label Printer, 165lb/75kg, 11.8" x 15.7" Platform 1113005 Mfr # BKT 165A
COMPARE
External 165 5
₹332,000.00INR / Each
Adam Equipment BKT 165AH Industrial Bench Scale with Built-In Label Printer, 165lb/75kg, 15.7 " x 19.7" Platform 1113006 Mfr # BKT 165AH
COMPARE
External calibration 165 2
Call For Price
Adam Equipment BKT 330A Industrial Bench Scale with Built-In Label Printer, 330lb/150kg, 15.7 " x 19.7"  Platform 1113007 Mfr # BKT 330A
COMPARE
External calibration 330 10
Call For Price
Adam Equipment BKT 330AH Industrial Bench Scale with Built-In Label Printer, 330lb/150kg, 15.7 " x 19.7"  Platform 1113008 Mfr # BKT 330AH
COMPARE
External calibration 330 5
Call For Price
Adam Equipment BKT 660A Industrial Bench Scale with Built-In Label Printer, 660lb/300kg, 15.7 " x 19.7"  Platform 1113009 Mfr # BKT 660A
COMPARE
External calibration 660 20
Call For Price
Adam Equipment BKT1320a Industrial Bench Scale with Built-In Label Printer, 1320lb/600kg, 21.6" x 31.5" Platform 1113010 Mfr # BKT 1320A
COMPARE
External 1320 50
₹443,000.00INR / Each
Showing 1- 11 of 11