Accessories for Thermo Scientific Tube Revolver

Expand the uses of your Thermo Scientific Tube Revolver with these optional accessories.
4,500.00 - ₹6,600.00INR / Pkg of 2

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific 88881005 Paddles for Tube Revolver, 10 to 15 mL; 2/Pk 0472571 Mfr # 88881005
COMPARE
Paddles for Tube Revolver, 10 to 15 mL; 2/Pk
₹4,500.00INR / Pkg of 2
Thermo Scientific 88881006 Paddles for Tube Revolver, 5 to 7 mL; 2/Pk 0472572 Mfr # 88881006
COMPARE
Paddles for Tube Revolver, 5 to 7 mL; 2/Pk
₹4,500.00INR / Pkg of 2
Thermo Scientific 88881007 Paddles for Tube Revolver, 1.5 to 2.0 mL; 2/Pk 0472573 Mfr # 88881007
COMPARE
Paddles for Tube Revolver, 1.5 to 2.0 mL; 2/Pk
₹4,500.00INR / Pkg of 2
Thermo Scientific 88881009 Paddle for Tube Revolver, 50 mL; 1/Pk 0472575 Mfr # 88881009
COMPARE
Paddle for Tube Revolver, 50 mL; 1/Pk
₹6,600.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat