Thermo Scientific 88881007 Paddles for Tube Revolver, 1.5 to 2.0 mL; 2/Pk
Representative image only.
Thermo Scientific 88881007 Paddles for Tube Revolver, 1.5 to 2.0 mL; 2/Pk
Thermo Scientific | Mfr: 88881007| Item: 0472573
Expand the uses of your Thermo Scientific Tube Revolver with these optional accessories.
₹4,500.00INR / Pkg of 2
₹4,500.00INR / Pkg of 2
Product Description
Key Features
  • 2 year warranty
More About this Item
Each paddle holds 15 x 1.5/2-mL tubes
Two year warranty
Specifications
  • Compatible WithThermo Scientific Tube Revolver
  • DescriptionPaddles for Tube Revolver, 1.5 to 2.0 mL; 2/Pk