A.P. Buck Inc. Bio-Culture™ Microbial Air Sampling Pumps
Quiet, compact microbial air sampler is perfect for unobstrusive indoor qir quality (IAQ) applications
  • Control flow rates from 30 to 120 L/min
  • Works with standard 90 mm x 15 mm petri dishes
  • Rechargeable battery runs up to 8 hours
  • Tripod mounting threads allows 90° sampling
  • Suitable for use in USP Sampling
Use this easy-to-use microbial air sampling pump to check the air quality within and around buildings to ensure the health and comfort of occupants. Air sampling pump with adjustable flow rate, rechargeable battery, and a built-in timer makes it simple to collect samples for analysis. Set pump to operate in "continuous" mode or use the built-in timer to take samples at timed intervals of 1, 2, 5, or 10 minutes.
Pump accepts a standard 90 x 15 mm petri dish for easy sample collection and analysis. Sampling head features 380 (1 mm diameter) holes and can be sterilized by autoclave or gas torch. Run pump up to 8 hours at 100 L/min using the rechargeable battery. The charger provides a battery recharge in about 3 hours and allows the unit to operate on AC power. Lit “complete” LED and three short beeps indicate end of sampling.
333,000.00 - ₹596,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
A.P. Buck Inc. Bio-Culture™ Microbial Air Sampling Pump; 230 VAC 0490091 Mfr # APB-708200
COMPARE
230 Bio-Culture™ Microbial Air Sampling Pump; 230 VAC
₹507,000.00INR / Each
A.P. Buck Inc. Bio-Culture™ Microbial Air Sampling Pump and Calibration Kit; 230 VAC 2100003 Mfr # APB-7082ZZ-KIT
COMPARE
230 Bio-Culture™ Microbial Air Sampling Pump and Calibration Kit; 230 VAC
Call For Price
A.P. Buck Inc. Bio-Culture™ Microbial Air Sampling Pump and Calibration Kit; 120 VAC 2100118 Mfr # APB-7080ZZ-KIT
COMPARE
120 Bio-Culture™ Microbial Air Sampling Pump and Calibration Kit; 120 VAC
₹596,000.00INR / Each
A.P. Buck Inc. Bio-Culture™ Microbial Air Sampling Pump; 120 VAC 2100119 Mfr # APB-7080ZZ
COMPARE
120 Bio-Culture™ Microbial Air Sampling Pump; 120 VAC
₹333,000.00INR / Each