A&D Weighing Moisture Balances
Four selectable heating modes allow greater control of temperature settings
  • Settings are clear and easy-to-view with vacuum fluorescent display (VFD)
  • Five selectable measurement programs
  • Self check functions
  • Progress window allows easy visibility
  • Safe and reliable design
These weighing moisture balances feature four heating modes: standard, quick, step, and famp operational mode. In addition, there are five selectable measurement programs: standard, quick, automatic, timer, and manual. The balances are easy to handle and the ergonomic design helps prevent burn injuries which improving workflow movements of samples. The large VFD display makes it easy to read setting values, change in moisture content, and see action status indicators.
Data interface is provided by the RS-232C interface and WinCT-moisture software. Data memory function allows for test results to be stored and recalled or outputted for additional analysis.
566,000.00 - ₹664,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Readability (g)
Availability Pricing
A&D Weighing ML-50 Moisture Determination Balance, 51g x 0.005g, 115 V 0104125 Mfr # ML-50
COMPARE
51 Halogen 0.005
₹566,000.00INR / Each
A&D Weighing MF-50 Moisture Determination Balance, 51g x 0.002g, 115 V 0104130 Mfr # MF-50
COMPARE
51 Halogen 0.002
₹664,000.00INR / Each
A&D Weighing MX-50 Moisture Determination Balance, 51g x 0.001g, 115 V 0104140 Mfr # MX-50
COMPARE
51 Halogen 0.001
Call For Price
A&D Weighing MS70 Moisture Determination Balance, 71g x 0.0001g, 115 V 0104145 Mfr # MS-70
COMPARE
71 Halogen 0.0001