A&D Weighing HW-CEP Series Industrial Bench Scales
Designed to weigh safely in hazardous environments
  • Ten-digit keypad simplifies setting comparator limits
  • Stainless steel pan allows for wet weighing and easy cleanup
  • Built-in key lock prevents unwanted changes
  • Large LCD with white backlight enables viewing in dim light
  • IP65-rated base for both dust and water
  • IECEx, ATEX, and FM/FMc certified
  • Auto shut off saves battery life
The HW-CEP industrial bench scales are specially designed to weigh safely in potentially explosive atmospheres and are approved for use in FM Class 1, Division 1, groups C & D hazardous areas. Red, green and amber LED comparator lights allow you to increase productivity by allowing fast check weighing.
Other features include a 10-digit keypad for setting comparator limits, a key lock function to prevent unwanted setting changes, auto-tare function, accumulation function which displays sum total of separate weighing results, counting function, comparator function, external calibration, and low battery indicator. Weighing modes include parts counting and accumulation function for totaling weigh data. Weighing units include kg, lb, oz, and lb-oz.
The scales operate on 4 D batteries. No need to secure power from a safe area via a safety barrier.
529,000.00 - ₹741,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Repeatability (g)
Availability Pricing
A&D Weighing HW-10KCEP Industrial Bench Scale, 20 lbs 1112700 Mfr # HW-10KCEP
COMPARE
10 20 2
₹529,000.00INR / Each
A&D Weighing HW-60KCEP Industrial Bench Scale, 150 lbs 1112701 Mfr # HW-60KCEP
COMPARE
60 150 10
₹595,000.00INR / Each
A&D Weighing HW-100KCEP Industrial Bench Scale, 200 lbs 1112702 Mfr # HW-100KCEP
COMPARE
100 200 20
₹689,000.00INR / Each
A&D Weighing HW-200KCEP Industrial Bench Scale, 500 lbs 1112703 Mfr # HW-200KCEP
COMPARE
220 500 40
₹741,000.00INR / Each