A&D Weighing HLWP Portable Washdown Scales
Durable waterproof scales for foodservice and general laboratory applications
  • Small footprint offers convenient placement anywhere on your benchtop
  • Durable stainless steel construction is ideal for foodservice applications and messy environments
  • Portable operation and auto-off feature conserve battery life
  • IP65/NEMA 4X-rated
  • Clear LCD offers excellent visibility of weighing results and 1⁄3000 high resolution
  • NSF-certified for use in foodservice
The HLWP series of stainless steel compact portion control scales are waterproof, accurate, and reliable. These durable digital scales are designed to withstand the rigors of daily use in a foodservice environment. Scales are IP65/NEMA 4X-rated. Clear LCD offers excellent visibility of weighing results and 1/3000 high resolution. All scales are NSF-certified for use in foodservice.
93,200.00 - ₹94,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing HL-300WPF Portion Control Washdown Scale, 300g x 0.1g 1111481 Mfr # HL-300WPF
COMPARE
0.3 0.66 115
₹94,900.00INR / Each
A&D Weighing HL-1000WPF Portion Control Washdown Scale, 1000g x 0.5g 1111482 Mfr # HL-1000WPF
COMPARE
1 2 115
₹94,900.00INR / Each
A&D Weighing HL-3000WPF Portion Control Washdown Scale, 3000g x 1g 1111483 Mfr # HL-3000WPF
COMPARE
3 6 115
Call For Price
A&D Weighing HL-3000WPNF Portion Control Washdown Scale, 3000g x 1g (NTEP) 1111484 Mfr # HL-3000WPNF
COMPARE
3 6 115
₹94,100.00INR / Each
A&D Weighing HL-3000LWPF Portion Control Washdown Scale, 3000g x 1g (Large Weigh Pan) 1111485 Mfr # HL-3000LWPF
COMPARE
3 6 115
₹93,200.00INR / Each
A&D Weighing HL-3000LWPNF Portion Control Washdown Scale, 3000g x 1g (NTEP) Large Weigh Pan 1111486 Mfr # HL-3000LWPNF
COMPARE
3 6 115
₹94,100.00INR / Each