A&D Weighing GX-A Apollo Gravimetric Dispense Bundles
Bundled system works in conjunction with digital pumps to give you what you need for high-accuracy gravimetric dispensing and filling
  • Maximize the accuracy of your dosing and filling applications with direct feedback and control of your pump
  • Compatible with drives with 25-pin (DB-25) remote I/O connector
  • Never lose data again — store weighing data in non-volatile memory
  • Complementary WinCT-Dispense software supports data collection and statistical analysis and simplifies set up and operation
  • Save power with the auto power-off function after 10 minutes of inactivity
  • Internal calibration saves time and money with quick calibration
  • Automatically self-calibrates when ambient temperature changes are detected
  • Calibrate on-demand with one-touch automatic calibration
  • Password-protection up to 11 users and 1 administrator allows for greater versitility while still protecting data
  • Large backlit LCD provides enhances visibility in poorly lit areas
681,000.00 - ₹1,074,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g)
Availability Pricing
A&D Weighing GX-203A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 220 g x 0.001 g 1111450 Mfr # GX-203A-GS
COMPARE
220 0.001
Call For Price
A&D Weighing GX-1603A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 1620 g x 0.001 g 1111451 Mfr # GX-1603A-GS
COMPARE
1620 0.001
₹1,009,000.00INR / Each
A&D Weighing GX-2002A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 2200 g x 0.01 g 1111452 Mfr # GX-2002A-GS
COMPARE
2200 0.01
₹681,000.00INR / Each
A&D Weighing GX-6002A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 6200 g x 0.01 g 1111453 Mfr # GX-6002A-GS
COMPARE
6200 0.01
₹902,000.00INR / Each
A&D Weighing GX-10002A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 10200 g x 0.01 g 1111454 Mfr # GX-10002A-GS
COMPARE
10200 0.01
₹1,074,000.00INR / Each
A&D Weighing GX-6001A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 6200 g x 0.1 g 1111455 Mfr # GX-6001A-GS
COMPARE
6200 0.1
₹694,000.00INR / Each
A&D Weighing GX-10001A-GS Apollo Gravimetric Dispensing Balance System, Internal Calibration; 10200 g x 0.1 g 1111456 Mfr # GX-10001A-GS
COMPARE
10200 0.1
₹928,000.00INR / Each
Showing 1- 7 of 7