A&D Weighing Galaxy HR-A(R) Analytical Balances
Exceptional features and a compact footprint at an affordable price
  • Fast stabilization in 2 seconds
  • Large, removable breeze break with antistatic coating
  • Rotary sliding doors requires no extra space at the rear of the balance
  • Counting mode with the Automatic Counting Accuracy Improvement (ACAI) function
  • Statistical calculation, comparator, below-balance weighing, and density determination functions
  • GLP/GMP/GCP/ISO-compliant output
  • Large reverse-backlit LCD
418,000.00 - ₹604,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing HR-100AZ Galaxy Analytical Balance, 102g x 0.1mg with Internal Calibration 1111113 Mfr # HR-100AZ
COMPARE
Internal 102
₹521,000.00INR / Each
A&D Weighing HR-150AZ Galaxy Analytical Balance, 152g x 0.1mg with Internal Calibration 1111114 Mfr # HR-150AZ
COMPARE
Internal 152
₹569,000.00INR / Each
A&D Weighing HR-250AZ Galaxy Analytical Balance, 252g x 0.1mg with Internal Calibration 1111115 Mfr # HR-250AZ
COMPARE
Internal 252
₹604,000.00INR / Each
A&D Weighing HR-100A Galaxy Analytical Balance, 102g x 0.1mg with External Calibration 1111116 Mfr # HR-100A
COMPARE
External 102
₹418,000.00INR / Each
A&D Weighing HR-150A Galaxy Analytical Balance, 152g x 0.1mg with External Calibration 1111117 Mfr # HR-150A
COMPARE
External 152
₹465,000.00INR / Each
A&D Weighing HR-250A Galaxy Analytical Balance, 252g x 0.1mgwith External Calibration 1111118 Mfr # HR-250A
COMPARE
External 252
₹507,000.00INR / Each