A&D Weighing FSi Washdown Industrial Scales
NEMA 4/IP65-rated for washdown use
  • 45-segment sweep LCD—quickly judge when target weight is approaching
  • Key in HI and LO set points, or recall them from up to 99 preset values
  • Large bright comparator lights in red, green, and yellow simplify repetitive check weighting
  • 100 item data memory
  • Stainless steel washdown construction
290,000.00 - ₹393,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing FS-15KIF Washdown Industrial Scale, 15kg (30lb) x 0.002lb 1111211 Mfr # FS-15KIF
COMPARE
15 30 115 / 230
₹290,000.00INR / Each
A&D Weighing FS-30KIF Washdown Industrial Scale, 30kg (70lb) x 0.005lb 1111221 Mfr # FS-30KIF
COMPARE
30 70 115 / 230
₹393,000.00INR / Each