A&D HT Portable Scales
Compact & portable scale with multiple weighing units
  • Battery or AC adapter operation
  • LCD display
  • Multiple weighing units
  • Storage/carrying case included
The HT-Series compact scales operate on 4 AA alkaline batteries or the optional 120VAC adapter. The handy storage and carrying case also accommodates storage for batteries. The HT-Series feature a low battery indicator, overload indication and a stable indicator for convenience.
25,900.00 - ₹50,200.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing HT-300 Compact Scales 310g x 0.1g Incl Carry Case WW-11111-29 Mfr # HT-300
COMPARE
External 310 0.1
₹25,900.00INR / Each
A&D Weighing HT-500 Compact Scales 510g x 0.1g Incl Carry Case WW-11111-30 Mfr # HT-500
COMPARE
External 510 0.1
₹31,800.00INR / Each
A&D Weighing HT-3000 Compact Scales 3100g x 1g Incl Carry Case WW-11111-31 Mfr # HT-3000
COMPARE
External 3100 1
₹25,900.00INR / Each
A&D Weighing HT-5000 Compact Scales 5100g x 1g Incl Carry Case WW-11111-32 Mfr # HT-5000
COMPARE
External 5100 1
₹31,800.00INR / Each
A&D Weighing HT-120 Compact Scales 120g x 0.01g Incl Carry Case WW-11115-77 Mfr # HT-120
COMPARE
External 120 0.01
₹50,200.00INR / Each