A&D EK-EP Intrinsically Safe Portable Balance
Weigh safely in potentially explosive areas
  • Intrinsically safe construction prevents spark generation
  • Compact, lightweight and portable—goes anywhere!
  • Durable 304 SS weighing pan is chemically resistant
  • Large backlit LCD is viewable in any lighting conditions
  • Comparator function provides fast accurate filling, packing, and sorting
  • Adjust response characteristics to compensate for drafts and vibrations
  • Use a cable lock with the built-in security ring to prevent theft
Featuring a compact and lightweight design and an easy-to-read LCD screen, these EK-EP Intrinsically Safe Weighing Scales are IECEx compliant, providing an effective weighing solution for a range of applications, including hazardous area weighing.
Highly portable, this IECEx scale can easily be taken on the go, making it a popular choice among the many IECEx weighing systems available. And because it’s an intrinsically safe weighing scale, you can trust that the top pan balance will provide precise weight measurement even in potentially explosive or hazardous environments and workplaces.
401,000.00 - ₹474,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pan Length (in) Pan Width (in) Capacity (g)
Availability Pricing
A&D Weighing EK-EP Instrinsically Safe Portable Balance, 300 g x 0.01 g 1113303 Mfr # EK-300AEP
COMPARE
4 3/8 4 3/8 300
₹401,000.00INR / Each
A&D Weighing EK-EP Instrinsically Safe Portable Balance, 3000 g x 0.1 g 1113304 Mfr # EK-3000AEP
COMPARE
6 5/8 5 1/4 3000
₹424,000.00INR / Each
A&D Weighing EK-EP Instrinsically Safe Portable Balance, 12,000 g x 1 g 1113305 Mfr # EK-12KAEP
COMPARE
6 5/8 5 1/4 12000
₹474,000.00INR / Each