YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable
Representative image only.
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable
YSI | Mfr: 627150-20| Item: 5935381
Get lab-quality results at the source
₹450,000.00INR / Each
₹450,000.00INR / Each
Buy It Together
Current Item
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable
5935381
₹450,000.00INR / Each
+
YSI Pro20i Dissolved Oxygen Meter, 1-m Cable, Polarographic
5935340
₹301,000.00INR / Each
+
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Only
5935370
₹331,000.00INR / Each
=
Total Price 1082000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Probes with cables 13-ft (4-m) to 100-ft (30-m) include cable management kit.
Key Features
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 20-m cable
  • 2 year warranty
More About this Item
Interchangeable digital "smart" probes store calibration data for a year, saving you time and calibration solution costs—no need to recalibrate if connected to different meters. Convenient cable management kits are included with probes featuring cable lengths from 13-ft (4-m) to 100-ft (30-m)
Two year warranty
Specifications
  • ApplicationsWater quality, waste water, natural resources, oxygen analysis
  • Min Temperature (° C)-5
  • Max Temperature (° C)70
  • Temperature CompensationYes
  • GuardStainless steel
  • ConnectorWaterproof
  • Cable Length (m)20
  • DescriptionOptical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable