YSI IDS pH sensor , Digital, Double Junction, 3m cable
Representative image only.
YSI IDS pH sensor , Digital, Double Junction, 3m cable
YSI | Mfr: 103741Y| Item: 0551050
Digital sensors for Plug-and-play capabilities
₹100,000.00INR / Each
₹100,000.00INR / Each
Buy It Together
Current Item
YSI IDS pH sensor , Digital, Double Junction, 3m cable
YSI IDS pH sensor , Digital, Double Junction, 3m cable
0551050
₹100,000.00INR / Each
+ =
Total Price 460000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quality IDS probes with 3-m cable
  • 1 year warranty
More About this Item
Intelligent Design Sensors (IDS) digitally store their serial number, unique ID, and calibration data. The sensors can be moved from one meter to another and transfer their calibration data. Just plug a sensor into a YSI Multilab meter to change the parameter to be measured.
YSI IDS digital pH sensor with plastic body, gel double Junction reference and, 3m cable.
1 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • Description, Digital, Double Junction, 3m cable