YSI 1FD350Y MultiLab Single-Channel Multi-parameter Benchtop Meter

Representative image only.

YSI 1FD350Y MultiLab Single-Channel Multi-parameter Benchtop Meter

YSI | Mfr: 1FD350Y| Item: 0551012

Maximum flexibility with IDS digital smart sensor recognition

₹360,000.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹360,000.00INR / Each

Buy It Together

Current Item

YSI 1FD350Y MultiLab Single-Channel Multi-parameter Benchtop Meter
YSI 1FD350Y MultiLab Single-Channel Multi-parameter Benchtop Meter
0551012

₹360,000.00INR / Each
+
YSI IDS pH sensor , Digital, Double Junction, 3m cable
0551050

₹100,000.00INR / Each
+ =
Total Price 484700.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

electrode stand, USB cable, and universal AC power adapter.

Key Features

  • Large easy-to-read graphical display with backlight
  • Plug-and-play sensors for pH, ORD, conductivity, resistivity, TDS, salinity, and optical DO/BOD
  • Digital sensor recognition, processing, and data transfer
  • Quality Sensor Control (QSC) system monitors the condition of pH electrodes
  • Continuous monitoring (CMC) function verifies sensor's measuring range against calibration range
  • Data logging with GLP time-and-date traceability and USB connectivity
  • 3 year warranty

More About this Item

YSI MultiLab line of one-, two-, and three-channel meters provide maximum flexibility with smart sensor recognition. The Intelligent Design Sensors (IDS) plug-and-play feature lets you decide the meter''s operation. Plug in a pH electrode and its a pH meter; replace with a conductivity probe and its a conductivity meter; same for oxygen--it is that easy!

The IP64-rated waterproof meters feature a large graphical display with backlight, manual and automatic data logging with GLP traceability, and convenient USB data transfer. Quality QSC and CMC functions ensure proper functioning of the electrode within the calibration range of meter.

The IDS electrodes digitally store their serial number, electrode type, unique ID, and calibration data. The sensors can also be moved from one meter to another and transfer their calibration data.

(Note-BNC electrode adapter does not work on single channel meter 05510-12.)

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001
  • pH Accuracy±0.004
  • Buffer RecognitionAutomatic
  • mV Resolution1 mV
  • mV Accuracy±1
  • Min Conductivity10 µS
  • Max Conductivity2000 mS
  • Conductivity Resolution0.1 µS
  • Conductivity Accuracy±0.5% of reading
  • Min TDS0
  • Max TDS1999 ppm
  • TDS Resolution0.1 ppt
  • Min Salinity0
  • Max Salinity70.0 ppt
  • Salinity Resolution0.1 ppt
  • Salinity Accuracy±0.5% of reading
  • Min Resistivity0.5 kΩ
  • Max Resistivity100 kΩ
  • Resistivity Resolution0.1 mΩ; 0.1 kΩ; 0.1 MΩ
  • Resistivity Accuracy±0.5% of reading
  • Min Dissolved Oxygen0
  • Max Dissolved Oxygen50 ppm / 500%
  • InterfaceUSB
  • WaterproofYes
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Width (cm)19.1
  • Height (cm)7.9
  • Width (in)7 1/2
  • Height (in)3 1/8
  • Probe IncludedNo
  • DescriptionMultiLab Single-Channel Multi-parameter Benchtop Meter
  • Warranty3 year
Reviews
Q&A