YSI IDS BOD sensor IDS Pro OBOD (optical BOD) Probe, 3 m cable
Representative image only.
YSI IDS BOD sensor IDS Pro OBOD (optical BOD) Probe, 3 m cable
YSI | Mfr: 626500| Item: 0551054
Digital sensors for Plug-and-play capabilities
₹313,000.00INR / Each
₹313,000.00INR / Each
Buy It Together
Current Item
YSI IDS BOD sensor IDS Pro OBOD (optical BOD) Probe, 3 m cable
YSI IDS BOD sensor IDS Pro OBOD (optical BOD) Probe, 3 m cable
0551054
₹313,000.00INR / Each
+ =
Total Price 673000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quality IDS probes with 3-m cable
  • 2 year warranty
More About this Item
Intelligent Design Sensors (IDS) digitally store their serial number, unique ID, and calibration data. The sensors can be moved from one meter to another and transfer their calibration data. Just plug a sensor into a YSI Multilab meter to change the parameter to be measured.
YSI IDS digital Pro OBOD sensor with optical BOD technology.
Two year warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • DescriptionIDS Pro OBOD (optical BOD) Probe, 3 m cable