YSI 301711Y IDS Conductivity Digital 4-Graphite Electrodes, 3m cable
Representative image only.
YSI 301711Y IDS Conductivity Digital 4-Graphite Electrodes, 3m cable
YSI | Mfr: 301711Y| Item: 0551058
Digital sensors for Plug-and-play capabilities
₹139,000.00INR / Each
₹139,000.00INR / Each
Buy It Together
Current Item
YSI 301711Y IDS Conductivity Digital 4-Graphite Electrodes, 3m cable
YSI 301711Y IDS Conductivity Digital 4-Graphite Electrodes, 3m cable
0551058
₹139,000.00INR / Each
+ =
Total Price 499000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quality IDS probes with 3-m cable
  • 1 year warranty
More About this Item
Intelligent Design Sensors (IDS) digitally store their serial number, unique ID, and calibration data. The sensors can be moved from one meter to another and transfer their calibration data. Just plug a sensor into a YSI Multilab meter to change the parameter to be measured.
YSI IDS digital conductivity sensor with epoxy body, 4-Graphite Electrode and, 3m cable.
1 Year Warranty
Specifications
  • Meter TypeAccessory; Probes and Sensors
  • DescriptionIDS Conductivity Digital 4-Graphite Electrodes, 3m cable