Wildco 1801-A12 Cover Slips for 05491-60; 12/Pk
Representative image only.
Wildco 1801-A12 Cover Slips for 05491-60; 12/Pk
Wildco | Mfr: 1801-A12| Item: 0549162
Easy counting of zooplankton
₹10,100.00INR / Each
₹10,100.00INR / Each
Buy It Together
Current Item
Wildco 1801-A12 Cover Slips for 05491-60; 12/Pk
Wildco 1801-A12 Cover Slips for 05491-60; 12/Pk
0549162
₹10,100.00INR / Each
+ =
Total Price 56700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Intended for use with standard or inverted compound microscopes at low magnifications
  • 3 month warranty
More About this Item
Coverslips for the Sedgewick-Rafter Counting Cell
3 Month Warranty
Specifications
  • MaterialGlass
  • Compatible WithFor 05491-60 Sedgewick-Rafter Counting Cell
  • Width (InchShort)1
  • DescriptionCover Slips for 05491-60; 12/Pk