W.S. Tyler 8537 Top Cover with Handle for 12" Diameter Stainless Steel Sieve
Representative image only.
W.S. Tyler 8537 Top Cover with Handle for 12" Diameter Stainless Steel Sieve
W.S. Tyler | Mfr: 8537| Item: 5999695
Protect and collect samples easily
₹17,100.00INR / Each
₹17,100.00INR / Each
Buy It Together
Current Item
W.S. Tyler 8537 Top Cover with Handle for 12" Diameter Stainless Steel Sieve
W.S. Tyler 8537 Top Cover with Handle for 12" Diameter Stainless Steel Sieve
5999695
₹17,100.00INR / Each
+ =
Total Price 64100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Order a top cover for a single or top
sieve to protect your sample from dust
contaminants. Order a receiving pan
for the bottom sieve to collect finest
sieved particles. Receiving pans with fitted
rim for nesting let you collect particles
between sieves.
Order a top cover when using a single sieve, or on the top sieve when stacked, to protect your sample from dust contaminants.
1 Year Warranty
Specifications
  • MaterialStainless Steel
  • Outside Diameter (in)12
  • Height (InchShort)0.5
  • DescriptionTop Cover with Handle for 12" Diameter Stainless Steel Sieve