W.S. Tyler 8534 Receiving Pan with Fitted Rim for Nesting 12" SS Full-Height Sieves
Representative image only.
W.S. Tyler 8534 Receiving Pan with Fitted Rim for Nesting 12" SS Full-Height Sieves
W.S. Tyler | Mfr: 8534| Item: 5999692
Protect and collect samples easily
₹36,600.00INR / Each
₹36,600.00INR / Each
Buy It Together
Current Item
W.S. Tyler 8534 Receiving Pan with Fitted Rim for Nesting 12" SS Full-Height Sieves
W.S. Tyler 8534 Receiving Pan with Fitted Rim for Nesting 12" SS Full-Height Sieves
5999692
₹36,600.00INR / Each
+ =
Total Price 83600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Order a top cover for a single or top
sieve to protect your sample from dust
contaminants. Order a receiving pan
for the bottom sieve to collect finest
sieved particles. Receiving pans with fitted
rim for nesting let you collect particles
between sieves.
Order a receiving pan for the bottom sieve to collect the finest particles. Receiving pans with a fitted rim for nesting allow you to collect from between sieves, not just the bottom sieve.
1 Year Warranty
Specifications
  • MaterialStainless Steel
  • Outside Diameter (in)12
  • Height (InchShort)3.25
  • DescriptionReceiving Pan with Fitted Rim for Nesting 12" SS Full-Height Sieves