W.S. Tyler 8533 Receiving Pan for 12" Stainless Steel Bottom Sieves, Half Height
Representative image only.
W.S. Tyler 8533 Receiving Pan for 12" Stainless Steel Bottom Sieves, Half Height
W.S. Tyler | Mfr: 8533| Item: 5999691
Protect and collect samples easily
₹31,200.00INR / Each
₹31,200.00INR / Each
Buy It Together
Current Item
W.S. Tyler 8533 Receiving Pan for 12" Stainless Steel Bottom Sieves, Half Height
W.S. Tyler 8533 Receiving Pan for 12" Stainless Steel Bottom Sieves, Half Height
5999691
₹31,200.00INR / Each
+ =
Total Price 78200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Order a top cover for a single or top
sieve to protect your sample from dust
contaminants. Order a receiving pan
for the bottom sieve to collect finest
sieved particles. Receiving pans with fitted
rim for nesting let you collect particles
between sieves.
Order a receiving pan for the bottom sieve to collect the finest particles. Receiving pans with a fitted rim for nesting allow you to collect from between sieves, not just the bottom sieve.
1 Year Warranty
Specifications
  • MaterialStainless Steel
  • Outside Diameter (in)12
  • Height (InchShort)1.625
  • DescriptionReceiving Pan for 12" Stainless Steel Bottom Sieves, Half Height