W.S. Tyler 8527 Receiving Pan for 12" Brass Bottom Sieves, Half Height

Representative image only.

W.S. Tyler 8527 Receiving Pan for 12" Brass Bottom Sieves, Half Height

W.S. Tyler | Mfr: 8527| Item: 5999681

Protect and collect samples easily

₹13,900.00INR / Each

₹13,900.00INR / Each

Buy It Together

Current Item

W.S. Tyler 8527 Receiving Pan for 12" Brass Bottom Sieves, Half Height
W.S. Tyler 8527 Receiving Pan for 12" Brass Bottom Sieves, Half Height
5999681

₹13,900.00INR / Each
+ =
Total Price 60900.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Order a top cover for a single or top
sieve to protect your sample from dust
contaminants. Order a receiving pan
for the bottom sieve to collect finest
sieved particles. Receiving pans with fitted
rim for nesting let you collect particles
between sieves.

Order a receiving pan for the bottom sieve to collect the finest particles. Receiving pans with a fitted rim for nesting allow you to collect from between sieves, not just the bottom sieve.

1 Year Warranty
Specifications
  • MaterialBrass
  • Outside Diameter (in)12
  • Height (InchShort)1.625
  • DescriptionReceiving Pan for 12" Brass Bottom Sieves, Half Height
  • Warranty1 Year
Reviews
Q&A