Luer fittings allow for a simple yet secure twist-lock connection
Ideal for capping off fluid transfer system
Prevent leaks with integral lock ring mechanism
Constructed of durable chemical resistance nylon
More About this Item
These Value Plastics Male Luer Integral Lock Ring to Plug Adapters are designed to easily plug female luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. These adapters are available in various materials to accommodate various applications and compliance requirements.
Choose from plugs closed at the tip and plugs closed at the grip. Plugs closed at luer tip are designed to seal at the tip of the plug, where it connects with the female luer fitting, ideal when it’s crucial to prevent any leakage or contamination at the tip of the fitting. Plugs closed at the grip feature a sealing mechanism located closer to the body of the plug, where it connects with the mating female luer fitting. This design may be preferred in applications where the tip of the fitting needs to remain open or where the focus is on sealing at the base rather than the tip.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
Fitting MaterialNylon
Width (InchShort)7/16
Height (InchShort)25/64
Number Of Ports1
Fitting Width (in)7/16
Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS
Luer Connection Type (Port 1)Male Luer Integral Lock Ring
Luer Connection Type (Port 2)Plug, Closed at Luer Tip
DescriptionLuer Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk