Cole-Parmer® Cleanroom Female Luer Plugs, Straight Adapter
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Forms a leak-free luer connection with a quick simple turn
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a male luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
800.00 - ₹4,700.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Material
Availability Pricing
COMPARE
Female Luer Plug CrystalVu™
₹2,600.00INR / Pkg of 10
COMPARE
Female Luer Plug Whie Nylon
₹4,700.00INR / Pkg of 10
COMPARE
Female Luer Plug Natural Kynar®
₹4,600.00INR / Pkg of 10
COMPARE
Female Luer Plug Natural Nylon
₹2,500.00INR / Pkg of 10
COMPARE
Female Luer Plug Polypropylene
₹2,500.00INR / Pkg of 10
COMPARE
Female Luer Plug Polypropylene
₹800.00INR / Pkg of 10