Value Plastics Luer Fitting, Straight Adapter, Transparent Polypropylene, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Straight Adapter, Transparent Polypropylene, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk
Value Plastics | Mfr: MTLLP-6005| Item: 5012075
Seal luer ports with a secure and leak-proof connection
₹2,500.00INR / Pkg of 25
₹2,500.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer Fitting, Straight Adapter, Transparent Polypropylene, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk
Value Plastics Luer Fitting, Straight Adapter, Transparent Polypropylene, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk
5012075
₹2,500.00INR / Pkg of 25
+ + =
Total Price 4890.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • Prevent leaks with integral lock ring mechanism
  • Constructed of ADCF polypropylene
More About this Item
These Value Plastics Male Luer Integral Lock Ring to Plug Adapters are designed to easily plug female luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. These adapters are available in various materials to accommodate various applications and compliance requirements.
Choose from plugs closed at the tip and plugs closed at the grip. Plugs closed at luer tip are designed to seal at the tip of the plug, where it connects with the female luer fitting, ideal when it’s crucial to prevent any leakage or contamination at the tip of the fitting. Plugs closed at the grip feature a sealing mechanism located closer to the body of the plug, where it connects with the mating female luer fitting. This design may be preferred in applications where the tip of the fitting needs to remain open or where the focus is on sealing at the base rather than the tip.
Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Width (InchShort)7/16
  • Height (InchShort)25/64
  • Number Of Ports1
  • Fitting Width (in)7/16
  • Regulatory ApprovalsFDA;Non-Phthalate;ADCF;RoHS;Non-DEHP;USP
  • Luer Connection Type (Port 1)Male Luer Integral Lock Ring
  • Luer Connection Type (Port 2)Plug, Closed at Luer Tip
  • DescriptionLuer Fitting, Straight Adapter, Transparent Polypropylene, Male Luer Integral Lock Ring x Plug, Closed at Luer Tip; 25/Pk