Value Plastics Hose Barb Fitting, Y Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
Representative image only.
Value Plastics Hose Barb Fitting, Y Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
Value Plastics | Mfr: Y230-6005| Item: 5012090
A secure connection under pressure with 200 series barbs
₹5,900.00INR / Pkg of 25
₹5,900.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Hose Barb Fitting, Y Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
Value Plastics Hose Barb Fitting, Y Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
5012090
₹5,900.00INR / Pkg of 25
+
C-Flex 072 TPE Tubing, 1/8" ID x 1/4" OD; 50 Ft
5012309
₹12,700.00INR / Pkg of 1
=
Total Price 18600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Constructed of USP compliant polypropylene
More About this Item
The Value Plastics 200 Series Y Hose Barb Fittings are designed to provide outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. The Y-orientation offers directional flexibility, while the barbs grip the inside of the hose, creating a tight seal to prevent leaks or disconnection.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Sterilization MethodGamma radiation, ethylene oxide (EtO), E-beam, and autoclave
  • Number Of Ports3
  • Connection TypeHose Barb Y
  • Barb Size / Tubing Id (Port 1) (mm)3.175
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Barb Size / Tubing Id (Port 2) (mm)3.175
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Barb Size / Tubing Id (Port 3) (mm)3.175
  • Barb Size / Tubing Id (Port 3) (in)1/8
  • Flow PatternY
  • ColorTransparent
  • Regulatory ApprovalsFDA;RoHS;USP;Non-DEHP;ADCF;Non-Phthalate
  • DescriptionHose Barb Fitting, Y Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk