C-Flex® 072 Thermoplastic Elastomer (TPE) Tubing
Excellent heat seal and sterile weld capabilities make it ideal for biopharmaceutical applications
  • Flexible and durable for use in peristaltic pumps
  • Less gas permeable compared to silicone
  • Opaque tubing—perfect for light sensitive fluids
  • USP Class VI and European Pharmacopeia 3.2.9 compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Easily meet the critical demands of pharmaceutical and biopharmaceutical applications with C-Flex TPE tubing. This moldable and bondable tubing is specifically designed for aseptic welding connections and sealing disconnection; seal or weld before or after sterilization depending on your application requirements.
Excellent flexibility and durability offer prolonged service life in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability characteristics protects sensitive fluids from gas ingress.
6,540.00 - ₹68,120.00INR / Pkg of 1
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
C-Flex 072 TPE Tubing, 1/32" ID x 3/32" OD; 50 Ft WW-50123-06 Mfr # 072-031-1
COMPARE
50 1/32 3/32
₹6,700.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/16" ID x1/8" OD; 50 Ft WW-50123-07 Mfr # 072-063-1
COMPARE
50 1/16 1/8
₹6,540.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/16" ID x 3/16" OD; 50 Ft WW-50123-08 Mfr # 072-063-2
COMPARE
50 1/16 3/16
₹10,420.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/8" ID x 1/4" OD; 50 Ft WW-50123-09 Mfr # 072-125-2
COMPARE
50 1/8 1/4
₹8,960.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/16" ID x 5/16" OD; 50 Ft WW-50123-10 Mfr # 072-188-2
COMPARE
50 3/16 5/16
₹9,020.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/16" ID x 3/8" OD; 50 Ft WW-50123-11 Mfr # 072-188-3
COMPARE
50 3/16 3/8
₹13,170.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/4" ID x 3/8" OD; 50 Ft WW-50123-12 Mfr # 072-250-2
COMPARE
50 1/4 3/8
₹13,500.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/4" ID x 7/16" OD; 50 Ft WW-50123-13 Mfr # 072-250-3
COMPARE
50 1/4 7/16
₹16,040.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/4" ID x 1/2" OD; 50 Ft WW-50123-14 Mfr # 072-250-4
COMPARE
50 1/4 1/2
₹20,730.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 5/16" ID x 1/2" OD; 50 Ft WW-50123-15 Mfr # 072-313-3
COMPARE
50 5/16 1/2
₹18,840.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/8" ID x 1/2" OD; 50 Ft WW-50123-16 Mfr # 072-375-2
COMPARE
50 3/8 1/2
₹17,110.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/8" ID x 9/16" OD; 50 Ft WW-50123-17 Mfr # 072-375-3
COMPARE
50 3/8 9/16
₹21,810.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/8" ID x 5/8" OD; 50 Ft WW-50123-18 Mfr # 072-375-4
COMPARE
50 3/8 5/8
₹28,610.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/2" ID x 11/16" OD; 50 Ft WW-50123-19 Mfr # 072-500-3
COMPARE
50 1/2 11/16
₹27,530.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1/2" ID x 3/4" OD; 50 Ft WW-50123-20 Mfr # 072-500-4
COMPARE
50 1/2 3/4
₹36,440.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 5/8" ID x 7/8" OD; 50 Ft WW-50123-21 Mfr # 072-625-4
COMPARE
50 5/8 7/8
₹68,120.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/4" ID x 1" OD; 15 Ft WW-50123-22 Mfr # 072-750-4
COMPARE
15 3/4 1
₹15,230.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/4" ID x 1-1/8" OD; 15 Ft WW-50123-23 Mfr # 072-750-6
COMPARE
15 3/4 1 1/8
₹26,610.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 3/4" ID x 1-1/4" OD; 15 Ft WW-50123-24 Mfr # 072-750-8
COMPARE
15 3/4 1 1/4
₹39,140.00INR / Pkg of 1
C-Flex 072 TPE Tubing, 1" ID x 1-3/8" OD; 15 Ft WW-50123-25 Mfr # 072-1000-6
COMPARE
15 1 1 3/8
₹41,460.00INR / Pkg of 1
Showing 1 - 20 of 21