United Scientific Supplies Short-Path Distillation Glassware Kit
Representative image only.
United Scientific Supplies Short-Path Distillation Glassware Kit
| Mfr: SPDK01| Item: 3450943
Compact design offers high production and consistent results
₹143,000.00INR / Each
₹143,000.00INR / Each
Buy It Together
Current Item
United Scientific Supplies Short-Path Distillation Glassware Kit
United Scientific Supplies Short-Path Distillation Glassware Kit
3450943
₹143,000.00INR / Each
+ =
Total Price 180200.00 ADD 2 ITEMS TO CART
Product Description
Includes
Short-path distillation head, 250-mL round-bottom two-neck flask, cow-type distillation receiver, glass inlet tube with tapered end, six plastic clamps, thermometer, three 50-mL single-neck round bottom flasks, rubber inlet adapter with glass connector, and three pennyhead glass stoppers.
Key Features
  • Made from 3.3 borosilicate glass for greater durability
  • Rubber inlet adapter safely holds tapered glass inlet tube
  • 1 year warranty
More About this Item
Manufactured from high-quality 3.3 borosilicate glass, this kit contains all the key glass components for short-path distillation. It features a short-path distillation head with 24/40 flask joints, a 10/30 thermometer port with a condenser coil, and three 1/4" hose nozzles (two for condenser and one for vacuum). Attach up to three 50-mL single-neck round-bottom flasks to the cow-type distillation receiver with one 24/40 outer joint and three 24/40 inner joints. A 250-mL round-bottom two-neck flask includes 24/40 outer joints to accept the distillation head and a rubber inlet adapter for the tapered glass inlet tube.
The thermometer, which connects to the distillation head through a 10/30 port, has a range of -10 to 50°C and is filled with a petroleum-based solution with red dye. Pennyhead glass stoppers are included to cap the single-neck round-bottom flasks after distillate is collected.
1 Year Warranty
Specifications
  • MaterialBorocilicate Glass
  • Capacity (mL)250
  • Joint Size Number24/40
  • DescriptionShort Path Distillation Kit