United Scientific Supplies Short-Path Distillation Head with Coil Condenser, 24/40 Joints
Representative image only.
United Scientific Supplies Short-Path Distillation Head with Coil Condenser, 24/40 Joints
| Mfr: DSH85| Item: 3450961
Ideal for botanical distillation or fragrance extraction
₹37,200.00INR / Each
₹37,200.00INR / Each
Product Description
Key Features
  • Made from 3.3 borosilicate glass for greater durability
  • Features three 1/4" hose nozzles
  • 1 year warranty
More About this Item
The short-path distillation head, manufactured from high-quality 3.3 borosilicate glass, features 24/40 flask joints with a 10/30 thermometer port. It also includes a condenser coil and three 1/4" hose nozzles (two for condenser and one for vacuum).
Short-path distillation is a distillation technique that involves the distillate travelling a short distance and done at reduced pressure. This process is ideal when higher temperatures of traditional distillation would degrade the sample to be distilled, for compounds that are unstable at high temperatures, or for purifying small amounts of compound.
1 Year Warranty
Specifications
  • MaterialGlass
  • Joint Size Number24/40
  • Tubing Sizes Accepted (in)1/4
  • DescriptionShort Path Distillation Head with Coil Condenser, 24/40 Joints