Type K PTFE-Coated Beaded Probe, Mini Connector
Representative image only.
Type K PTFE-Coated Beaded Probe, Mini Connector
| Mfr: 4028CC| Item: 9876721
Easily connect to any type K themocouple thermometer
₹3,900.00INR / Each
₹3,900.00INR / Each
Buy It Together
Current Item
Type K PTFE-Coated Beaded Probe, Mini Connector
Type K PTFE-Coated Beaded Probe, Mini Connector
9876721
₹3,900.00INR / Each
+ +
Traceable Infrared Stick Thermometer; 8:1
2025009
₹25,500.00INR / Each
+ + =
Total Price 111300.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Mini connectors provide quick connections to type K thermometers
  • Replacement probe for Traceable® meters 37803-91, 37803-94, 91210-01, 91210-03, and 91210-30
  • 1 year warranty
More About this Item
These probes are available in many types such as beaded, penetration, and with or without a handle.
For improved performance, probes should be calibrated as a system with the meter.
Maximum temperature rating is 572°F (300°C) for short-term use.
1 Year Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-40
  • Max Temperature (° F)482
  • Min Temperature (° C)-40
  • Max Temperature (° C)250
  • Connector TypeMini Connector
  • Sheath MaterialPTFE coated
  • Sheath Length (in)48
  • Sheath Diameter (InchShort)0.06
  • Cable TypePTFE coated
  • Cable Length (InchShort)48
  • DescriptionType K PTFE-Coated Beaded Probe, Mini Connector