Troemner 974037 Wall Bracket with secure message and safety chain, Gas Cylinder Clamp
Representative image only.
Troemner 974037 Wall Bracket with secure message and safety chain, Gas Cylinder Clamp
Troemner | Mfr: 974037| Item: 0812968
₹34,200.00INR / Each
₹34,200.00INR / Each
Buy It Together
Current Item
Troemner 974037 Wall Bracket with secure message and safety chain, Gas Cylinder Clamp
Troemner 974037 Wall Bracket with secure message and safety chain, Gas Cylinder Clamp
0812968
₹34,200.00INR / Each
+
Troemner 971046 Safety Strap with Secure Message
0812959
₹15,300.00INR / Each
=
Total Price 49500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 2 year warranty
More About this Item
Secure gas cylinders to increase safety. Straps feature "Secure Cylinder" text that is easy to read when the gas cylinder is properly strapped into place.
Two year warranty
Specifications
  • Clamp MaterialCast Aluminum and Nylon
  • Clamp TypeEquipment, Gas Cylinder
  • Min Grip Range (InchShort)4
  • Max Grip Range (InchShort)14
  • DescriptionWall Bracket with secure message and safety chain, Gas Cylinder Clamp