Troemner 971048 Safety Strap without Message
Representative image only.
Troemner 971048 Safety Strap without Message
Troemner | Mfr: 971048| Item: 0812962
₹6,600.00INR / Each
₹6,600.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Secure gas cylinders to increase safety.
Two year warranty
Specifications
  • Clamp MaterialNylon and Nickel-plated Zinc
  • Clamp TypeSafety Strap without Message
  • Min Grip Range (InchShort)4
  • Max Grip Range (InchShort)14
  • Width (in)1
  • Length (in)54
  • DescriptionSafety Strap without Message