Troemner 974036 Bench Clamp with secure message and safety chain, Gas Cylinder Clam
Representative image only.
Troemner 974036 Bench Clamp with secure message and safety chain, Gas Cylinder Clam
Troemner | Mfr: 974036| Item: 0812967
₹36,200.00INR / Each
₹36,200.00INR / Each
Buy It Together
Current Item
Troemner 974036 Bench Clamp with secure message and safety chain, Gas Cylinder Clam
Troemner 974036 Bench Clamp with secure message and safety chain, Gas Cylinder Clam
0812967
₹36,200.00INR / Each
+
Troemner 970035 Heavy Duty Bench Clamp (Cast Aluminum)
0812954
₹28,200.00INR / Each
=
Total Price 64400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 2 year warranty
More About this Item
Secure gas cylinders to increase safety. Straps feature "Secure Cylinder" text that is easy to read when the gas cylinder is properly strapped into place.
Two year warranty
Specifications
  • Clamp MaterialCast Aluminum and Nylon
  • Clamp TypeEquipment, Gas Cylinder
  • Min Grip Range (InchShort)4
  • Max Grip Range (InchShort)14
  • DescriptionBench Clamp with secure message and safety chain, Gas Cylinder Clam