Troemner 973036 Bench Clamp with secure message, Gas Cylinder Clamp
Representative image only.
Troemner 973036 Bench Clamp with secure message, Gas Cylinder Clamp
Troemner | Mfr: 973036| Item: 0812965
₹32,800.00INR / Each
₹32,800.00INR / Each
Buy It Together
Current Item
Troemner 973036 Bench Clamp with secure message, Gas Cylinder Clamp
Troemner 973036 Bench Clamp with secure message, Gas Cylinder Clamp
0812965
₹32,800.00INR / Each
+
Troemner 970035 Heavy Duty Bench Clamp (Cast Aluminum)
0812954
₹28,200.00INR / Each
=
Total Price 61000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 2 year warranty
More About this Item
Secure gas cylinders to increase safety. Straps feature "Secure Cylinder" text that is easy to read when the gas cylinder is properly strapped into place.
Two year warranty
Specifications
  • Clamp MaterialCast Aluminum and Nylon
  • Clamp TypeEquipment, Gas Cylinder
  • Min Grip Range (InchShort)4
  • Max Grip Range (InchShort)14
  • DescriptionBench Clamp with secure message, Gas Cylinder Clamp