Troemner 9078T 25 Lb Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate
Representative image only.
Troemner 9078T 25 Lb Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate
Troemner | Mfr: 9078T| Item: 1196654
Acceptable for use verifying scalse that have a ±0.1% accuracy or lower
₹49,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹49,400.00INR / Each
Buy It Together
Current Item
Troemner 9078T 25 Lb Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate
Troemner 9078T 25 Lb Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate
1196654
₹49,400.00INR / Each
+ =
Total Price 66900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Color coding – silver for avoirdupois
  • 3 month warranty
More About this Item
Cast Iron Grip Handle Weights larger than 5 kg/10 lb are legal for trade, which means their design is acceptable for use in checking weighing devices where the price of the item being purchased is based on its weight.
Cast Iron Grip Handle Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Cast Iron Grip Handle weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable for use in verifying scales that have a 0.1% accuracy and lower.
3 Month Warranty
Specifications
  • Mass ClassClass F
  • Mass Size (lbs)25
  • Mass CertificationNIST
  • Description25 lb, Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate