Troemner 7028-0W 200 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate

Representative image only.

Troemner 7028-0W 200 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate

Troemner | Mfr: 7028-0W| Item: 0102105

UltraClass - The weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2. This class is used as a reference standard for calibrating other reference standards and is appropriate for calibrating highprecision analytical balances with a readability as low as 0.01 mg.

₹38,700.00INR / Each

₹38,700.00INR / Each

Buy It Together

Current Item

Troemner 7028-0W 200 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate
Troemner 7028-0W 200 mg, UltraClass Analytical Mass with NVLAP Accredited Certificate
0102105

₹38,700.00INR / Each
+ +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
+
Environmental Express Buffer Solution, pH 12.46; 500 mL
0065412

₹2,100.00INR / Each
+ + + + + + =
Total Price 482800.00 ADD 10 ITEMS TO CART
Product Description

Includes

a clear-top polycarbonate case, all weights include noted documentation of accuracy from the vendor.

Key Features

  • UltraClass Tolerance = ASTM Class 0
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel

More About this Item

These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes. The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerancesdecrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.

Specifications
  • Mass ClassUltraClass
  • Mass Size (mg)200
  • Mass Size (g)0.2
  • Mass Size (lbs)0.00044
  • Mass CertificationNVLAP
  • Tolerance0.005 mg
  • Height (in)0.0937
  • Width (in)0.3281
  • Length (in)0.2968
  • Description200 mg, Analytical Ultraclass Mass with NVLAP Accredited Certificate
Reviews
Q&A