Trajan SGE Dispenser Syringe for Hamilton C and CX, 250 µL
Representative image only.
Trajan SGE Dispenser Syringe for Hamilton C and CX, 250 µL
Trajan | Mfr: 006995| Item: 1600134
Precision-crafted for performance and reliability, Trajan SGE Dispenser Syringes are engineered to meet the demands of analytical and laboratory automation, ensuring seamless integration and dependable results
₹12,300.00INR / Each
₹12,300.00INR / Each
Buy It Together
Current Item
Trajan SGE Dispenser Syringe for Hamilton C and CX, 250 µL
Trajan SGE Dispenser Syringe for Hamilton C and CX, 250 µL
1600134
₹12,300.00INR / Each
+ + + +
B-D Disposable Syringe, Non-Sterile, Luer-Lok, 20 mL; 25/Pk
0794416
₹4,500.00INR / Pkg of 25
=
Total Price 96700.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • High accuracy and precision
  • Robust, durable design minimizes downtime
  • Inert fluid path using borosilicate glass and premium PTFE
  • Seamlessly replaces Hamilton C, CX, V3, and V6 pump syringes
  • Easy to install with standard thread fittings
  • 6 month warranty
More About this Item
Trajan SGE Dispenser Syringes are built to deliver consistent performance in automated systems and laboratory applications. With a focus on precision, durability, and chemical inertness, these syringes feature borosilicate glass barrels and PTFE fluid paths to minimize interaction with samples. Designed as drop-in replacements for Hamilton C, CX, V3, and V6 syringes, they offer easy installation, a wide range of capacities, and compatibility with standard 1/4-28 UNF fittings. Their robust construction helps reduce maintenance downtime, making them a reliable and efficient choice for precision-driven workflows.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Gas Tight Syringe, Compatible with PerkinElmer Autosampler, 250 uL; 1/Ea