Hamilton® Gastight® Syringes with PTFE Luer Lock
Gas and liquid-tight up to 100 psi
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Sample loss is negligible at pressures up to 100 psi
Hamilton® Gastight® syringes with PTFE luer lock connections are constructed with a chemically inert glass barrel. Gastight syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze. Needles are not included, order separately.
9,100.00 - ₹65,200.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Syringe Capacity (mL) Description
Availability Pricing
Hamilton Gastight® Syringe with PTFE Luer Lock, 100 mL, 100 psig WW-07938-33 Mfr # 86020
COMPARE
Borosilicate glass, PTFE, PCTFE 100 Syringe with PTFE Luer Lock, 100 mL, 100 psig
₹65,200.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 50 µL, 1000 psig WW-07939-32 Mfr # 80920
COMPARE
Borosilicate glass, PTFE 0.05 Syringe with PTFE Luer Lock, 50 µL, 1000 psig
₹9,100.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 100 µL, 1000 psig WW-07939-33 Mfr # 81020
COMPARE
Borosilicate glass, PTFE 0.1 Syringe with PTFE Luer Lock, 100 µL, 1000 psig
₹15,000.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 250 µL, 500 psig WW-07939-34 Mfr # 81120
COMPARE
Borosilicate glass, PTFE 0.25 Syringe with PTFE Luer Lock, 250 µL, 500 psig
₹15,000.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 500 µL, 700 psig WW-07939-35 Mfr # 81220
COMPARE
Borosilicate glass, PTFE 0.5 Syringe with PTFE Luer Lock, 500 µL, 700 psig
₹10,000.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 1 mL, 200 psig WW-07939-80 Mfr # 81320
COMPARE
Borosilicate glass, PTFE 1 Syringe with PTFE Luer Lock, 1 mL, 200 psig
₹11,300.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 2.5 mL, 200 psig WW-07939-82 Mfr # 81420
COMPARE
Borosilicate glass, PTFE 2.5 Syringe with PTFE Luer Lock, 2.5 mL, 200 psig
₹12,800.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 5 mL, 200 psig WW-07939-83 Mfr # 81520
COMPARE
Borosilicate glass, PTFE 5 Syringe with PTFE Luer Lock, 5 mL, 200 psig
₹13,900.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 10 mL, 200 psig WW-07939-84 Mfr # 81620
COMPARE
Borosilicate glass, PTFE 10 Syringe with PTFE Luer Lock, 10 mL, 200 psig
₹16,500.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 25 mL, 100 psig WW-07939-85 Mfr # 82520
COMPARE
Borosilicate glass, PTFE, PCTFE 25 Syringe with PTFE Luer Lock, 25 mL, 100 psig
₹44,600.00INR / Each
Hamilton Gastight® Syringe with PTFE Luer Lock, 50 mL, 100 psig WW-07939-86 Mfr # 85020
COMPARE
Borosilicate glass, PTFE, PCTFE 50 Syringe with PTFE Luer Lock, 50 mL, 100 psig
₹54,900.00INR / Each
Showing 1- 11 of 11