Trajan SGE 031842 Replacement Plunger, for 1 mL Gas Tight Syringe, Removable Bevel-Tip Needle
Representative image only.
Trajan SGE 031842 Replacement Plunger, for 1 mL Gas Tight Syringe, Removable Bevel-Tip Needle
Trajan | Mfr: 031842| Item: 1600263
Choose wisely to save your operational cost—no need to buy new syringes instead
₹7,600.00INR / Pkg of 1
₹7,600.00INR / Pkg of 1
Buy It Together
Current Item
Trajan SGE 031842 Replacement Plunger, for 1 mL Gas Tight Syringe, Removable Bevel-Tip Needle
Trajan SGE 031842 Replacement Plunger, for 1 mL Gas Tight Syringe, Removable Bevel-Tip Needle
1600263
₹7,600.00INR / Pkg of 1
+
Trajan SGE Dispenser Syringe for Hamilton C and CX, 250 µL
1600134
₹12,300.00INR / Each
=
Total Price 19900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Plunger, for 1 mL Gas Tight Syringe, Removable BT Needle; 1/Ea