Traceable Waterproof Temperature Data Logger Kit
Representative image only.
Traceable Waterproof Temperature Data Logger Kit
Traceable | Mfr: 6541TR| Item: 1800501
Comes with everything you to download and analyze your temperature data
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Traceable Waterproof Temperature Data Logger Kit
Traceable Waterproof Temperature Data Logger Kit
1800501
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Includes
Traceable® Certificate, five data loggers, one copy of data download and analysis software, one cradle with USB cable, and battery.
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Compact and reusable—ideal for transportation monitoring
  • Download data and prepare reports with thorough data analysis
  • IP66-rated housing for use in virtually any environment
  • Stores 7,680 temperature data points with time and date stamp
  • Only measures 2.56" x 1.34" x 0.35" and weighs 0.65 ounces
  • 1 year warranty
More About this Item
The kit comes with five data loggers, software, and cradle with USB cable—everything you need to quickly download and analyze from multiple loggers. One copy of the software may be used for multiple loggers and allows you to update settings including individual unit name/description, time zone settings, daylight savings time on/off, unit of measure (°F or °C), start delay, recording duration/sampling interval, quarantine/alarm settings, and user stop key enabling.
Each compact, lightweight data logger is reusable and reprogrammable making it ideal for transportation monitoring. Unit comes preprogrammed with alarm setpoints at 2°C (low) and 8°C (high) and to set record temperature readings every 5 minutes and 33 seconds for 30 days of continuous monitoring. For complete flexibility, the user-defined recording interval may be set based on the number of desired days to log (from 4 hours to 365 days) or the desired time interval (from every 2 seconds to every 24 hours). If the measurement falls outside of the user-determined alarm conditions for too long, "QUARANTINE" is shown on the display—perfect for monitoring environmental temperatures of refrigerated vaccines, chemicals, pharmaceuticals, and other perishable items.
The IP66 waterproof rating allows the units to be used in any location, even those that are damp or require washdown. Order the optional sleeves to provide other users with a quick reference of the logger settings including minimum, maximum, quarantine, logging interval and duration, start and stop date and time, and special instructions.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
IP 66 RatedNIST Traceable Calibration1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature
  • Max Temperature (° F)122
  • Min Temperature (° F)-31
  • Max Temperature (° C)50
  • Min Temperature (° C)-35
  • Temperature Resolution (° F)0.1
  • Temperature Accuracy (° F)±0.4
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.2
  • Memory Size7680 temperature readings with time and date stamp
  • Alarm0 to 99 for "QUARANTINE"
  • Battery Type1 3V CR2450 lithium (included)
  • Display TypeLCD
  • DescriptionWaterproof Temperature Data Logger Kit