Traceable 7600 Smart Temperature Bottle Probe
Representative image only.
Traceable 7600 Smart Temperature Bottle Probe
Traceable | Mfr: 7652TR| Item: 0750412
Save time and money by simply replacing probes that are damaged or due for calibration
₹17,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹17,000.00INR / Each
Buy It Together
Current Item
Traceable 7600 Smart Temperature Bottle Probe
Traceable 7600 Smart Temperature Bottle Probe
0750412
₹17,000.00INR / Each
+ + =
Total Price 96700.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Thermometer recalibration accomplished by simply replacing the probe
  • Durable 10-ft (3-m) long microcable
  • The 7600-series data logger will auto recognize the probes’ parameters
  • Three calibration points -40, 0, and 50°C
  • A door gasket can be closed on the probe cables for use in refrigerators and freezers
  • Bottle filled with nontoxic glycol solution (GRAS) generally recognized as safe by the FDA
  • Temperature-buffered glycol bottle probe mimics solution
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
Calibration data are stored in the probe, so simply replace the probe when it is time for recalibration. Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
7600-Series Smart Temperature Probes are calibrated in place of the base data logger. They remove downtime in the lab by allowing an outdated sensor to be quickly swapped out with a new pre-calibrated one—no need to worry about sending the data logger for recalibration. Outdated sensors can then be disposed of or recalibrated without losing any valuable time.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
Specifications
  • Probe TypeBottle
  • Min Temperature (° F)-58
  • Max Temperature (° F)140
  • Min Temperature (° C)-50
  • Max Temperature (° C)60
  • Cable TypeMicrocable
  • Cable Length (InchShort)108
  • Compatible WithWi-Fi Data Logger compatible with Traceable 7600 Smart Probes and TraceableLIVE® Cloud Service
  • DescriptionSmart Temperature Bottle Probe