Traceable 6600 Ethernet Smart Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service

Representative image only.

Traceable 6600 Ethernet Smart Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service

Traceable | Mfr: 6600TR| Item: 9946001

Stay connected to your critical environments—wherever you go!

₹46,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹46,700.00INR / Each

Buy It Together

Current Item

Traceable 6600 Ethernet Smart Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service
Traceable 6600 Ethernet Smart Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service
9946001

₹46,700.00INR / Each
+
Traceable 7600 Smart Temperature Bottle Probe
0750412

₹17,000.00INR / Each
+ + =
Total Price 111600.00 ADD 4 ITEMS TO CART
Product Description

Includes

Traceable® Certificate, one bottle probe with 10-ft (3-m) long cable, holder with magnetic back, Ethernet cable, universal power adapter and plug with 5-ft (1.5-m) long cord, and batteries.

Key Features

  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE cloud service*
  • Get the reliability, speed, and security using Ethernet connection
  • View current temperature conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
  • Includes one glass bottle probe
  • Bottle filled with nontoxic glycol solution (GRAS) generally recognized as safe by the FDA
  • Temperature-buffered glycol bottle probe mimics solution
  • Use to measure temperatures ranging from -50 to 60°C (-58 to 140°F)
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

TraceableLIVE Wireless Technology allows multiple users to stay connected and monitor critical environments 24/7 wherever you go.

These data logging thermometers with Ethernet offer an easy and secure way to connect your smartphone, tablet or personal computer remotely with your water baths, refrigerators, freezers and biological/pharmaceutical lab equipment.

Once setup is complete, you can securely monitor parameters 24/7, control your alarms remotely, view your data logging history, run reports in real time and give access to other team members. Stay connected with your lab via smartphone, tablet or computer, with no local software is required to use the cloud-based interface. Get the TraceableLIVE app to view mobile push notifications, emails, or texts for out-of-range conditions or connectivity interruptions. Large tripe-line LCD displays current and Min/Max readings. The thermometer features audible and visual alarms help those present in the lab identify any out-of-range conditions. A low-cost subscription is required for TraceableLIVE use.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

*Requires a TraceableLIVE subscription.

Conformite EuropeenneNIST Traceable Calibration1 Year Warranty
Specifications
  • Number of Channels1
  • Cloud ConnectedYes
  • Input TypeBottle Probe
  • Max Temperature (° F)140
  • Min Temperature (° F)-58
  • Min Temperature (° C)-50
  • Max Temperature (° C)60
  • Temperature Resolution (° F)0.01
  • Temperature Accuracy (° F)±0.18
  • Temperature Resolution (° C)0.01
  • Temperature Accuracy (° C)±0.10
  • Sampling Rate6 seconds
  • Memory Size672 on unit; infinite on cloud
  • InterfaceEthernet
  • System Requirementsweb, Android™, or Apple® iOS
  • AlarmYes
  • ConnectionsEthernet
  • Battery Type3 AAA (included)
  • Display TypeLCD
  • Length (in)2 3/4
  • Height (in)5 1/2
  • Depth (in)1
  • Length (cm)7
  • Height (cm)14
  • Depth (cm)2.5
  • DescriptionEthernet Smart Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service
  • Warranty1 year
Reviews
Q&A