ThermoSafe 440KD Corrugated Carton for Utility Mailers item number 03741-20

Representative image only.

ThermoSafe 440KD Corrugated Carton for Utility Mailers item number 03741-20

ThermoSafe | Mfr: 440KD| Item: 0374123

Ship nonhazardous specimens safely

₹12,800.00INR / Case of 24

₹12,800.00INR / Case of 24

Buy It Together

Current Item

ThermoSafe 440KD Corrugated Carton for Utility Mailers item number 03741-20
ThermoSafe 440KD Corrugated Carton for Utility Mailers item number 03741-20
0374123

₹12,800.00INR / Case of 24
+
ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs
0372701

₹22,600.00INR / Case of 150
=
Total Price 35400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Shipping cartons secure your mailers

More About this Item

White, cardboard, shipping sleeves cover the foam test tube mailers securely and safely to help prevent damage to the insulation—get multiple uses out of your shipping systems! Foam mailers are sold separately.

Specifications
  • Shipping Weight (oz)149
  • Shipping Weight (g)4218
  • DescriptionShipping Carton for 03741-20
Reviews
Q&A