ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs

Representative image only.

ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs

ThermoSafe | Mfr: 339| Item: 0372701

Ship non-hazardous specimens safely

₹22,600.00INR / Case of 150

₹22,600.00INR / Case of 150

Buy It Together

Current Item

ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs
ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs
0372701

₹22,600.00INR / Case of 150
+
ThermoSafe 365 Shipping Container/Carton for 03727-01
0372723

₹27,000.00INR / Case of 300
=
Total Price 49600.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

More About this Item

Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.

Specifications
  • CapacityFive 13 x 103 mm tubes
  • Inside Length (in)4 3/16
  • Outside Width (in)4 1/2
  • Outside Length (in)5 1/8
  • Outside Depth (in)1 1/2
  • Inside Diameter (ID) (in)9/16
  • Inside Length (cm)10.6
  • Outside Width (cm)11.4
  • Outside Length (cm)13
  • Outside Depth (cm)3.8
  • Inside Diameter (ID) (cm)1.4
  • Shipping Weight (oz)64
  • Shipping Weight (g)1814
  • DescriptionShipper, 5 Tubes, 13 x 103 mm tubes, 150/cs
Reviews
Q&A