ThermoSafe 365 Shipping Container/Carton for 03727-01

Representative image only.

ThermoSafe 365 Shipping Container/Carton for 03727-01

ThermoSafe | Mfr: 365| Item: 0372723

Ship nonhazardous specimens safely

₹27,000.00INR / Case of 300

₹27,000.00INR / Case of 300

Buy It Together

Current Item

ThermoSafe 365 Shipping Container/Carton for 03727-01
ThermoSafe 365 Shipping Container/Carton for 03727-01
0372723

₹27,000.00INR / Case of 300
+
ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs
0372701

₹22,600.00INR / Case of 150
=
Total Price 49600.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Shipping cartons secure your mailers

More About this Item

White, cardboard, shipping sleeves cover the foam test tube mailers securely and safely to help prevent damage to the insulation—get multiple uses out of your shipping systems! Foam mailers are sold separately.

Specifications
  • Shipping Weight (oz)271
  • Shipping Weight (g)7688
  • DescriptionShipping Carton for 03727-01
Reviews
Q&A