ThermoSafe 376 Dry Ice Storage Chest Dolly, fits 3.75 cu ft chest
Representative image only.
ThermoSafe 376 Dry Ice Storage Chest Dolly, fits 3.75 cu ft chest
ThermoSafe | Mfr: 376| Item: 0372010
Easily transport your large-capacity dry ice storage chests
₹55,200.00INR / Each
₹55,200.00INR / Each
Buy It Together
Current Item
ThermoSafe 376 Dry Ice Storage Chest Dolly, fits 3.75 cu ft chest
ThermoSafe 376 Dry Ice Storage Chest Dolly, fits 3.75 cu ft chest
0372010
₹55,200.00INR / Each
+ =
Total Price 302200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Large 4" (10.2 cm) swivel casters roll easily
  • Constructed of durable aluminum for reliable transportation
  • NSF certified
More About this Item
Move your large-capacity storage chests easily on these aluminum dollies, even when filled to capacity with dry ice. The rim on the edge of platform keeps containers on the dolly during acceleration, bumps, or stops. Dollys feature a gray hammertone finish on heavy-gauge aluminum. Storage chests sold separately.
Specifications
  • MaterialAluminum
  • Compatible WithStorage Chest 03720-00
  • DescriptionDry Ice Storage Chest Dolly, fits 3.75 cu ft chest