ThermoSafe 373 Boston Round Bottle Shipper, Vertical, 4 oz/118 mL; 100/Cs

Representative image only.

ThermoSafe 373 Boston Round Bottle Shipper, Vertical, 4 oz/118 mL; 100/Cs

ThermoSafe | Mfr: 373| Item: 0373132

Ship nonhazardous specimens safely

₹27,100.00INR / Case of 100

₹27,100.00INR / Case of 100

Buy It Together

Current Item

ThermoSafe 373 Boston Round Bottle Shipper, Vertical, 4 oz/118 mL; 100/Cs
ThermoSafe 373 Boston Round Bottle Shipper, Vertical, 4 oz/118 mL; 100/Cs
0373132

₹27,100.00INR / Case of 100
+
ThermoSafe 339 Shipper, 5 Tube Mailer, 13 x 103 mm tubes, 150/cs
0372701

₹22,600.00INR / Case of 150
=
Total Price 49700.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Shipping cartons secure your mailers

More About this Item

White, cardboard, shipping sleeves cover the foam test tube mailers securely and safely to help prevent damage to the insulation—get multiple uses out of your shipping systems! Foam mailers are sold separately.

Specifications
  • Bottle Capacity (mL)118
  • Inside Height (in)6 1/4
  • Inside Length (in)5 1/4
  • Inside Width (in)3 1/8
  • Outside Width (in)3 1/8
  • Outside Length (in)3 1/8
  • Outside Height (in)6 1/4
  • Inside Height (cm)15.9
  • Inside Length (cm)13.3
  • Inside Width (cm)7.9
  • Outside Width (cm)7.9
  • Outside Length (cm)7.9
  • Outside Height (cm)15.9
  • Shipping Weight (oz)96
  • Shipping Weight (g)2721
  • DescriptionShipping Carton for 03727-04
Reviews
Q&A