ThermoProbe TP9A-050-SW-SM Stainless Steel 50ft Std Probe

Representative image only.

ThermoProbe TP9A-050-SW-SM Stainless Steel 50ft Std Probe

ThermoProbe | Mfr: TP9A-050-SW-SM| Item: 9002604

Intrinsically safe thermometer for accurate temperature measurement in bulk liquid storage tanks

₹570,000.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹570,000.00INR / Each

Buy It Together

Current Item

ThermoProbe TP9A-050-SW-SM Stainless Steel 50ft Std Probe
ThermoProbe TP9A-050-SW-SM Stainless Steel 50ft Std Probe
9002604

₹570,000.00INR / Each
+ +
ThermoProbe PG-050-SW-SM Replacement Probe Assembly 50ft Std
9002606

₹166,000.00INR / Each
=
Total Price 780700.00 ADD 3 ITEMS TO CART
Product Description

Includes

2 AA batteries and NIST traceable calibration report supplied by manufacturer.

Key Features

  • Ideal alternative to hazardous glass thermometers
  • Reliable and stable RTD sensor technology
  • Accuracy ±0.2°F from 32 to 200°F (0 to 100°C)
  • Easy to calibrate and make adjustments using faceplate controls
  • Cable marked every 5 feet
  • Includes 4-point NIST traceable calibration report
  • 1 year warranty

More About this Item

The ThermoProbe are designed as rugged, field-use, portable precision digital thermometers, involving volume-temperature gauging of petroleum and chemical products. Convenient design allows for easy retrieval of the cable probe and eliminates the need to bring the probe to the surface to take multiple readings. Typical applications include custody transfers, tanks, pipelines, ships, railcars, barges, tanker trucks, and proving systems.

Zone 0 intrinsically safe meters are manufactured of materials which are both immune to petrochemicals and are nonsparking. The probe assembly is constructed with a very flexible, static-dissipating, aramid fiber reinforced, nonstick cable and stainless steel components.

Large LCD displays temperature, plus arrows indicate the direction of the temperature reading and whether stability has been reached. Stabilized temperatures can be logged at numerous liquid levels for a running average and later displayed for documenting. Advanced features, accessible through a user-friendly menu, include Min/Max/Avg reading, readout integrity, photosensor-controlled backlight, selectable (°F/°C) unit of measure, battery status, and automatic shut-off.

Model TP9A features a durable, corrosion-resistant stainless steel case and a wraparound design for retrieval of the cable with probe.

Conformite EuropeenneIntrinsically Safe1 Year Warranty
Specifications
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-40
  • Max Temperature (° F)400
  • Min Temperature (° C)-40
  • Max Temperature (° C)204
  • Accuracy±0.2°F; ±0.5°F above 200°F [±0.1°C; ±0.3°C above 100°C]
  • Resolution0.01°
  • Low Battery IndicatorYes
  • Height (in)10
  • No. of Probe(s) Included1
  • Battery Life200 hours
  • Battery2 AA (included)
  • DescriptionStainless Steel 50ft Std Probe
  • Warranty1 year
Reviews
Q&A