Thermo Scientific TSG General Purpose Chromatography Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC

Representative image only.

Thermo Scientific TSG General Purpose Chromatography Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC

Thermo Scientific | Mfr: TSG49CSGA| Item: 4426158

Go green while improving cold storage

₹4,128,000.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹4,128,000.00INR / Each

Buy It Together

Current Item

Thermo Scientific TSG General Purpose Chromatography Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC
Thermo Scientific TSG General Purpose Chromatography Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC
4426158

₹4,128,000.00INR / Each
+ + =
Total Price 4128000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

adjustable shelves and half shelves, shelving clips, door lock key, power key switch, and power cord.

Key Features

  • EPA ENERGY STAR® certified high-efficiency cooling system
  • Green, natural refrigerants offer greater safety
  • Whisper-quiet compressor and low heat output fits in any lab setting
  • Temperature high and low, door ajar, and power failure alarms protect critical supplies and samples
  • Protect samples with the safety sensor interior outlet
  • Feature a painted gray exterior and stainless steel interior, 49 cubic foot capacity, and double glass doors

More About this Item

Ideal for storing reagents, media, and samples, these general purpose chromatography refrigerators are also engineered to be better for the environment. Save electricity and money with adaptive defrost, decreased compressor cycle time, and reduced heat transfer. The natural refrigerant R290 will not deplete the ozone layer or contribute substantially to climate change. Being manufactured in a zero-waste-to-landfill facility ensures at least 99% of the waste produced is reused, recycled, composted, incinerated, or used in energy recovery.

The refrigerators’ quiet 51-dBA operation and low heat output allows for installation near workstations for added convenience. Visible and audible alarms alert you when temperature exceeds high or low limits and when there is a power outage prevent losses of valuable contents. Durable steel construction withstands chemical spills and resists corrosion.

The chromatography refrigerators are equipped with a hydrocarbon safety circuit sensor to ensure safety. In the event the safety circuit is tripped and the safety sensor senses a flammable gas, the power to the internal outlet will be discontinued.

UL Listed to Canadian safety standards.Two year warranty
Specifications
  • Refrigerator Capacity (cu ft)49
  • Refrigerator Capacity (Liters)1387
  • Min Temperature - Refrigerator (° C)2
  • Max Temperature - Refrigerator (° C)10
  • Temperature ControlDigital
  • Refrigerator Interior Width (in)28 1/2
  • Refrigerator Interior Height (in)49
  • Refrigerator Interior Depth (in)61 3/4
  • Refrigerator Interior Width (cm)72.4
  • Refrigerator Interior Height (cm)124.5
  • Refrigerator Interior Depth (cm)157
  • Exterior Width (in)32
  • Exterior Height (in)52 45/64
  • Exterior Depth (in)81
  • Exterior Width (cm)81.3
  • Exterior Height (cm)134
  • Exterior Depth (cm)205.7
  • Number of Doors2
  • Number of Shelves8 full, 2 half
  • AlarmYes
  • Cabinet ConstructionStainless Steel
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)8.8
  • DescriptionGeneral Purpose Chromatography Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC
Reviews
Q&A