Thermo Scientific SH480X1 Ceramic Shelf, for 350 cu-ft furnaces
Representative image only.
Thermo Scientific SH480X1 Ceramic Shelf, for 350 cu-ft furnaces
Thermo Scientific | Mfr: SH480X1| Item: 0205361
₹27,400.00INR / Each
₹27,400.00INR / Each
Buy It Together
Current Item
Thermo Scientific SH480X1 Ceramic Shelf, for 350 cu-ft furnaces
Thermo Scientific SH480X1 Ceramic Shelf, for 350 cu-ft furnaces
0205361
₹27,400.00INR / Each
+ + + =
Total Price 2098400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Ceramic hearth plates, shelves and other parts and accessories for your muffle furnace.
Specifications
  • Capacity (cu in)350
  • Max Temperature (° C)1200
  • DescriptionCeramic Shelf, for 350 cu-ft furnaces